missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, 97%

Catalog No. AAA1923803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1923803 1 g
AAA1923806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1923803 Supplier Thermo Scientific Chemicals Supplier No. A1923803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1632-19-5 | C8H9NO6 | 215.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1632-19-5
Molecular Formula C8H9NO6
Molecular Weight (g/mol) 215.161
MDL Number MFCD00276587
InChI Key QCPBGHYMHYWRHG-UHFFFAOYSA-N
Synonym dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, maybridge4_003392, acmc-20al76, 2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 2,5-dimethyl 3,4-dihydroxy-1h-pyrrole-2,5-dicarboxylate, methyl 3,4-dihydroxy-5-methoxycarbonyl pyrrole-2-carboxylate, 2z,5z-2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 1h-pyrrole-2,5-dicarboxylicacid, 3,4-dihydroxy-, 2,5-dimethyl ester
PubChem CID 2727733
IUPAC Name dimethyl 3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate
SMILES COC(=O)C1=C(C(=C(N1)C(=O)OC)O)O

Specifications

Melting Point 209°C to 212°C
Density 1.248
Boiling Point 268°C to 272°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 215.16
Percent Purity 97%
Chemical Name or Material Dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only