missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 3,4-Dihydroxy-2,5-thiophenedicarboxylate 98.0+%, TCI America™
$161.72 - $567.92
Chemical Identifiers
| CAS | 58416-04-9 |
|---|---|
| Molecular Formula | C8H8O6S |
| Molecular Weight (g/mol) | 232.206 |
| MDL Number | MFCD01860087 |
| InChI Key | ZZVINKJCOXPIKL-UHFFFAOYSA-N |
| Synonym | 3,4-Dihydroxy-2,5-thiophenedicarboxylic Acid Dimethyl Ester |
| PubChem CID | 350052 |
| IUPAC Name | dimethyl 3,4-dihydroxythiophene-2,5-dicarboxylate |
| SMILES | COC(=O)C1=C(C(=C(S1)C(=O)OC)O)O |
Chemical Identifiers
| 58416-04-9 | |
| 232.206 | |
| ZZVINKJCOXPIKL-UHFFFAOYSA-N | |
| 350052 | |
| COC(=O)C1=C(C(=C(S1)C(=O)OC)O)O |
| C8H8O6S | |
| MFCD01860087 | |
| 3,4-Dihydroxy-2,5-thiophenedicarboxylic Acid Dimethyl Ester | |
| dimethyl 3,4-dihydroxythiophene-2,5-dicarboxylate |
Specifications
| 58416-04-9 | |
| White-Yellow | |
| MFCD01860087 | |
| 3,4-Dihydroxy-2,5-thiophenedicarboxylic Acid Dimethyl Ester | |
| COC(=O)C1=C(C(=C(S1)C(=O)OC)O)O | |
| 232.206 | |
| 232.21 | |
| Crystalline Powder |
| 182°C | |
| C8H8O6S | |
| 1 g | |
| ZZVINKJCOXPIKL-UHFFFAOYSA-N | |
| dimethyl 3,4-dihydroxythiophene-2,5-dicarboxylate | |
| 350052 | |
| ≥98.0% (GC) | |
| Dimethyl 3,4-Dihydroxy-2,5-thiophenedicarboxylate |
Safety and Handling
TSCA : No