missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 3,4-Dihydroxy-2,5-thiophenedicarboxylate 98.0+%, TCI America™

Catalog No. D36825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36825G 5 g
D36821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36825G Supplier TCI America Supplier No. D36825G
Only null left

Chemical Identifiers

CAS 58416-04-9
Molecular Formula C8H8O6S
Molecular Weight (g/mol) 232.206
MDL Number MFCD01860087
InChI Key ZZVINKJCOXPIKL-UHFFFAOYSA-N
Synonym 3,4-Dihydroxy-2,5-thiophenedicarboxylic Acid Dimethyl Ester
PubChem CID 350052
IUPAC Name dimethyl 3,4-dihydroxythiophene-2,5-dicarboxylate
SMILES COC(=O)C1=C(C(=C(S1)C(=O)OC)O)O

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 5 g
Formula Weight 232.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 3,4-Dihydroxy-2,5-thiophenedicarboxylate