missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2-Oxopropyl)phosphonate 95.0+%, TCI America™

Catalog No. o020825g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
O020825G 25 g
O02085G 5 g
2 options

Catalog No. O020825G

Supplier: TCI America O020825G

Only null left

Chemical Identifiers

CAS 4202-14-6
Molecular Formula C5H11O4P
Molecular Weight (g/mol) 166.113
MDL Number MFCD00008769
InChI Key UOWIYNWMROWVDG-UHFFFAOYSA-N
Synonym dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
PubChem CID 77872
IUPAC Name 1-dimethoxyphosphorylpropan-2-one
SMILES CC(=O)CP(=O)(OC)OC

Specifications

Color Yellow
Boiling Point 124°C
Quantity 25 g
Formula Weight 166.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (2-Oxopropyl)phosphonate
TSCA No
Recommended Storage Refrigerator