missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2-Oxo-3-phenoxypropyl)phosphonate 98.0+%, TCI America™

Catalog No. D44081G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44081G 1 g
D4408200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44081G Supplier TCI America Supplier No. D44081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40665-68-7
Molecular Formula C11H15O5P
Molecular Weight (g/mol) 258.21
MDL Number MFCD00010225
InChI Key NQTSTBMCCAVWOS-UHFFFAOYSA-N
Synonym (2-Oxo-3-phenoxypropyl)phosphonic Acid Dimethyl Ester, Dimethyl (3-Phenoxyacetonyl)phosphonate, (3-Phenoxyacetonyl)phosphonic Acid Dimethyl Ester
PubChem CID 580104
IUPAC Name dimethyl (2-oxo-3-phenoxypropyl)phosphonate
SMILES COP(=O)(CC(=O)COC1=CC=CC=C1)OC

Specifications

Color Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 258.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (2-Oxo-3-phenoxypropyl)phosphonate