missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl [2-Oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate 98.0+%, TCI America™

Catalog No. d43971g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D43971G Supplier TCI America Supplier No. D43971G
Only null left

Chemical Identifiers

CAS 54094-19-8
Molecular Formula C12H14F3O5P
Molecular Weight (g/mol) 326.208
MDL Number MFCD09054713
InChI Key MBGWNNJXMYHKQO-UHFFFAOYSA-N
Synonym [2-Oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonic Acid Dimethyl Ester
PubChem CID 15003092
IUPAC Name 1-dimethoxyphosphoryl-3-[3-(trifluoromethyl)phenoxy]propan-2-one
SMILES COP(=O)(CC(=O)COC1=CC=CC(=C1)C(F)(F)F)OC

Specifications

Quantity 1 g
Formula Weight 326.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl [2-Oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate