missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,6-pyridinedicarboxylate, 99%

Catalog No. AC427940500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427940500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427940500 Supplier Thermo Scientific Chemicals Supplier No. 427940500
Only null left

CAS: 5453-67-8 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5453-67-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00134493
InChI Key SNQQJEJPJMXYTR-UHFFFAOYSA-N
Synonym dimethyl 2,6-pyridinedicarboxylate, 2,6-dimethyl pyridine-2,6-dicarboxylate, pyridine-2,6-dicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid, dimethyl ester, dimethyl dipicolinate, dimethyl pyridine-2,6-carboxylate, chembl72405, dimethylpyridine-2,6-dicarboxylate, dipicolinic acid dimethyl ester
PubChem CID 79549
IUPAC Name dimethyl pyridine-2,6-dicarboxylate
SMILES COC(=O)C1=NC(=CC=C1)C(=O)OC

Specifications

Melting Point 123°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Formula Weight 195.17
Percent Purity 99%
Chemical Name or Material Dimethyl 2, 6-pyridinedicarboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 226-697-3

RUO – Research Use Only