missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (+)-2,3-O-Isopropylidene-D-tartrate 95.0+%, TCI America™

Catalog No. I04745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04745G Supplier TCI America Supplier No. I04745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37031-30-4
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.205
MDL Number MFCD00040504
InChI Key ROZOUYVVWUTPNG-WDSKDSINSA-N
Synonym 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate
PubChem CID 688156
IUPAC Name dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
SMILES CC1(OC(C(O1)C(=O)OC)C(=O)OC)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 218.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (+)-2,3-O-Isopropylidene-D-tartrate