missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%

CAS: 37031-30-4 | C9H14O6 | 218.21 g/mol

$302.19 - $302.19

Chemical Identifiers

CAS 37031-30-4
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
MDL Number MFCD00040504
InChI Key ROZOUYVVWUTPNG-WDSKDSINSA-N
Synonym 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate
PubChem CID 688156
IUPAC Name dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
SMILES CC1(OC(C(O1)C(=O)OC)C(=O)OC)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC258440100
SDS View Documents
Thermo Scientific Chemicals
258440100
10 mL Glass bottle
Each for $302.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37031-30-4
218.21
ROZOUYVVWUTPNG-WDSKDSINSA-N
688156
CC1(OC(C(O1)C(=O)OC)C(=O)OC)C
C9H14O6
MFCD00040504
4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate
dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate

Specifications

37031-30-4
100.0
1.1500g/mL
135°C
96% min. (GC)
C9H14O6
MFCD00040504
1.15
+ 54
ROZOUYVVWUTPNG-WDSKDSINSA-N
dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
688156
97%
(+)-Dimethyl 2, 3-O-isopropylidene-D-tartrate
96.0
Colorless to Yellow
139°C (21.0 hPa)
Authentic
Glass bottle
1.4386 to 1.4406
10 mL
+ 54.00 (20.00°C c=neat)
4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate
CC1(OC(C(O1)C(=O)OC)C(=O)OC)C
218.21
218.21
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only