missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%

Catalog No. AC258440100
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC258440100 10 mL Glass bottle
AC258440500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC258440100 Supplier Thermo Scientific Chemicals Supplier No. 258440100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37031-30-4 | C9H14O6 | 218.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37031-30-4
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
MDL Number MFCD00040504
InChI Key ROZOUYVVWUTPNG-WDSKDSINSA-N
Synonym 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate
PubChem CID 688156
IUPAC Name dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
SMILES CC1(OC(C(O1)C(=O)OC)C(=O)OC)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 139°C (21.0 hPa)
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4386 to 1.4406
Quantity 10 mL
Specific Gravity 1.15
Specific Rotation Condition + 54.00 (20.00°C c=neat)
Specific Rotation + 54
Formula Weight 218.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (+)-Dimethyl 2, 3-O-isopropylidene-D-tartrate
Show More Show Less

RUO – Research Use Only