missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (1-Diazo-2-oxopropyl)phosphonate 97.0+%, TCI America™

Catalog No. D35461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35461G 1 g
D35465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35461G Supplier TCI America Supplier No. D35461G

Chemical Identifiers

CAS 90965-06-3
Molecular Formula C5H9N2O4P
Molecular Weight (g/mol) 192.111
MDL Number MFCD07368360
InChI Key SQHSJJGGWYIFCD-UHFFFAOYSA-N
Synonym (1-Diazo-2-oxopropyl)phosphonic Acid Dimethyl Ester, Dimethyl 1-Diazoacetonylphosphonate, 1-Diazoacetonylphosphonic Acid Dimethyl Ester, Ohira-Bestmann Reagent
PubChem CID 53397103
IUPAC Name 1-diazonio-1-dimethoxyphosphorylprop-1-en-2-olate
SMILES CC(=C([N+]#N)P(=O)(OC)OC)[O-]

Specifications

Quantity 1 g
Formula Weight 192.11
Percent Purity ≥97.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Dimethyl (1-Diazo-2-oxopropyl)phosphonate
TSCA No
Recommended Storage Refrigerator