missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-Phenylenediacrylate 95.0+%, TCI America™

Catalog No. P13385G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13385G Supplier TCI America Supplier No. P13385G
Only null left

Chemical Identifiers

CAS 7549-44-2
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.26
MDL Number MFCD00185690
InChI Key IFNSXAJHSAPYLB-UHFFFAOYSA-N
Synonym 1,4-Phenylenediacrylic Acid Dimethyl Ester
PubChem CID 6025991
IUPAC Name methyl 3-[4-(3-methoxy-3-oxoprop-1-en-1-yl)phenyl]prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(C=CC(=O)OC)C=C1

Specifications

Melting Point 167°C
Color Yellow
Quantity 5 g
Formula Weight 246.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 1,4-Phenylenediacrylate