missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%

Catalog No. p-7027335
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2400822 100 g
AAB2400836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2400822 Supplier Thermo Scientific Chemicals Supplier No. B2400822
Only null left

CAS: 6289-46-9 | C10H12O6 | 228.20 g/mol

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.

Solubility
Soluble in water 0.0016 g/L.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6289-46-9
Molecular Formula C10H12O6
Molecular Weight (g/mol) 228.20
MDL Number MFCD00001607
InChI Key MHKKFFHWMKEBDW-UHFFFAOYNA-N
Synonym dimethyl succinylsuccinate, dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, succinosuccinic acid, dimethyl ester, dmss, dimethyl succinylosuccinate, dimethyl 2,5-dioxo-1,4-cyclohexanedicarboxylate, dimethyl succinylo succinate, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, dimethyl ester, 2,5-dioxo-1,4-cyclohexanedicarboxylic acid dimethyl ester, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, 1,4-dimethyl ester
PubChem CID 94866
SMILES COC(=O)C1CC(=O)C(CC1=O)C(=O)OC

Specifications

Melting Point 152°C to 156°C
Quantity 100 g
Solubility Information Soluble in water 0.0016g/L.
IUPAC Name dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Formula Weight 228.2
Percent Purity ≥99%
Chemical Name or Material Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-528-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only