missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 98+%

Catalog No. AC271815000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271815000 500 g Glass bottle
AC271811000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC271815000 Supplier Thermo Scientific Chemicals Supplier No. 271815000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6289-46-9 | C10H12O6 | 228.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6289-46-9
Molecular Formula C10H12O6
Molecular Weight (g/mol) 228.20
MDL Number MFCD00001607
InChI Key MHKKFFHWMKEBDW-UHFFFAOYNA-N
Synonym dimethyl succinylsuccinate, dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, succinosuccinic acid, dimethyl ester, dmss, dimethyl succinylosuccinate, dimethyl 2,5-dioxo-1,4-cyclohexanedicarboxylate, dimethyl succinylo succinate, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, dimethyl ester, 2,5-dioxo-1,4-cyclohexanedicarboxylic acid dimethyl ester, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, 1,4-dimethyl ester
PubChem CID 94866
IUPAC Name dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
SMILES COC(=O)C1CC(=O)C(CC1=O)C(=O)OC

Specifications

Melting Point 154°C to 157°C
Color Green-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 500 g
Beilstein 10, IV, 3514
Formula Weight 228.2
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Dimethyl 1, 4-cyclohexanedione-2, 5-dicarboxylate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-528-9
TSCA TSCA

RUO – Research Use Only