missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 1,3-Benzodithiol-2-ylphosphonate 98.0+%, TCI America™
Supplier: TCI America D39921G
Specifications
Dimethyl 1,3-Benzodithiol-2-ylphosphonate | |
122°C | |
C9H11O3PS2 | |
1,3-Benzodithiol-2-ylphosphonic Acid Dimethyl Ester, 2-Dimethoxyphosphinyl-1,3-benzodithiole | |
COP(=O)(C1SC2=CC=CC=C2S1)OC | |
262.278 | |
262.28 | |
Crystalline Powder |
62217-35-0 | |
White-Yellow | |
1 g | |
VKVOUPYVLWQXLB-UHFFFAOYSA-N | |
2-dimethoxyphosphoryl-1,3-benzodithiole | |
400748 | |
≥98.0% (GC) |
Chemical Identifiers
62217-35-0 | |
262.278 | |
1,3-Benzodithiol-2-ylphosphonic Acid Dimethyl Ester, 2-Dimethoxyphosphinyl-1,3-benzodithioleShow More | |
2-dimethoxyphosphoryl-1,3-benzodithiole |
C9H11O3PS2 | |
VKVOUPYVLWQXLB-UHFFFAOYSA-N | |
400748 | |
COP(=O)(C1SC2=CC=CC=C2S1)OC |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator