missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethoxypillar[5]arene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 249°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 750.89 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dimethoxypillar[5]arene |
Safety and Handling
| TSCA | No |