missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxypillar[5]arene 98.0+%, TCI America™

Catalog No. D4471100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4471100MG 100 mg
D44711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4471100MG Supplier TCI America Supplier No. D4471100MG
Only null left

Chemical Identifiers

CAS 1188423-16-6
Molecular Formula C45H50O10
Molecular Weight (g/mol) 750.89
MDL Number MFCD28386103
InChI Key HJDHAGKELBPBLT-UHFFFAOYSA-N
Synonym DMpillar[5]arene
PubChem CID 50940580
IUPAC Name 4,9,14,20,25,27,29,30,32,34-decamethoxyhexacyclo[21.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹]pentatriaconta-1(25),3,5,8,10,13,15,18,20,23,26,28,30,32,34-pentadecaene
SMILES COC1=CC2=C(OC)C=C1CC1=CC(OC)=C(CC3=CC(OC)=C(CC4=C(OC)C=C(CC5=C(OC)C=C(C2)C(OC)=C5)C(OC)=C4)C=C3OC)C=C1OC

Specifications

Melting Point 249°C
Color Yellow
Quantity 100 mg
Formula Weight 750.89
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethoxypillar[5]arene