missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethylvinylsilane, 97%

Catalog No. AAL1403514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1403514 25 g
AAL1403522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1403514 Supplier Thermo Scientific Chemicals Supplier No. L1403514
Only null left

CAS: 16753-62-1 | C5H12O2Si | 132.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16753-62-1
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00129170
InChI Key ZLNAFSPCNATQPQ-UHFFFAOYSA-N
Synonym vinylmethyldimethoxysilane, dimethoxymethylvinylsilane, dimethoxy methyl vinyl silane, silane, ethenyldimethoxymethyl, ethenyldimethoxymethylsilane, methyldimethoxyvinylsilane, vinyl methyldimethoxysilane, dimethoxy methyl vinylsilane #, methylvinyldimethoxysilane
PubChem CID 85589
IUPAC Name ethenyl-dimethoxy-methylsilane
SMILES CO[Si](C)(C=C)OC

Specifications

Boiling Point 102°C to 104°C
Quantity 25 g
UN Number UN1993
Beilstein 3043765
Flash Point 5°C (41°F)
Formula Weight 132.23
Percent Purity 97%
Refractive Index 1.395
Sensitivity Moisture sensitive
Density 0.889
Chemical Name or Material Dimethoxymethylvinylsilane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 240-816-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only