missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethyl(3,3,3-trifluoropropyl)silane, 97%

Catalog No. p-7047025
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAL1729206 5 g
AAL1729214 25 g
2 options

Catalog No. AAL1729206

Supplier: Thermo Scientific Chemicals L1729206

Only null left

CAS: 358-67-8 | C6H13F3O2Si | 202.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 358-67-8
Molecular Formula C6H13F3O2Si
Molecular Weight (g/mol) 202.248
MDL Number MFCD00053175
InChI Key DIJRHOZMLZRNLM-UHFFFAOYSA-N
Synonym dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl
PubChem CID 67751
IUPAC Name dimethoxy-methyl-(3,3,3-trifluoropropyl)silane
SMILES CO[Si](C)(CCC(F)(F)F)OC

Specifications

Boiling Point 85°C (150mmHg)
Quantity 5 g
UN Number UN1993
Beilstein 1753237
Flash Point 58°C (136°F)
Formula Weight 202.25
Percent Purity 97%
Refractive Index 1.3576
Sensitivity Moisture sensitive
Density 1.1
Chemical Name or Material Dimethoxymethyl(3,3,3-trifluoropropyl)silane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-619-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only