missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane 98.0+%, TCI America™

Catalog No. D063725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D063725ML 25 mL
D0637500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D063725ML Supplier TCI America Supplier No. D063725ML

Chemical Identifiers

CAS 109-87-5
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00008495
InChI Key NKDDWNXOKDWJAK-UHFFFAOYSA-N
Synonym methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
PubChem CID 8020
ChEBI CHEBI:48341
IUPAC Name dimethoxymethane
SMILES COCOC

Specifications

Color Colorless
Boiling Point 42°C
Quantity 25 mL
UN Number 1234
Formula Weight 76.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxymethane
EINECSNumber (2)-0498
RTECSNumber PA8750000
TSCA Yes