missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydiphenylsilane 98.0+%, TCI America™

Catalog No. d1731500ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
D1731500ML 500 mL
D173125ML 25 mL
D1731100ML 100 mL
3 options

Catalog No. D1731500ML

Supplier: TCI America D1731500ML

Only null left

Chemical Identifiers

CAS 6843-66-9
Molecular Formula C14H16O2Si
Molecular Weight (g/mol) 244.365
MDL Number MFCD00025690
InChI Key AHUXYBVKTIBBJW-UHFFFAOYSA-N
Synonym diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s
PubChem CID 81284
IUPAC Name dimethoxy(diphenyl)silane
SMILES CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC

Specifications

Color Colorless
Boiling Point 110°C
Quantity 500 mL
Formula Weight 244.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxydiphenylsilane
EINECSNumber (3)-2635
RTECSNumber VV3643332
TSCA Yes