missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydiphenylsilane 98.0+%, TCI America™

Catalog No. D173125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D173125ML 25 mL
D1731100ML 100 mL
D1731500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D173125ML Supplier TCI America Supplier No. D173125ML

Chemical Identifiers

CAS 6843-66-9
Molecular Formula C14H16O2Si
Molecular Weight (g/mol) 244.365
MDL Number MFCD00025690
InChI Key AHUXYBVKTIBBJW-UHFFFAOYSA-N
Synonym diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s
PubChem CID 81284
IUPAC Name dimethoxy(diphenyl)silane
SMILES CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC

Specifications

Color Colorless
Boiling Point 110°C
Quantity 25 mL
Formula Weight 244.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxydiphenylsilane
EINECSNumber (3)-2635
RTECSNumber VV3643332
TSCA Yes