Learn More
Dimethoxydiphenylsilane, 97%
CAS: 6843-66-9 | C14H16O2Si | 244.365 g/mol
Supplier: Thermo Scientific Chemicals L040310B
Description
Reagent for preparation of diphenylsilylene derivatives of diols by exchange.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 160°C to 162°C (15mmHg) | |
| 6843-66-9 | |
| C14H16O2Si | |
| 2940458 | |
| diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s | |
| CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC | |
| 244.365 | |
| 244.37 | |
| 1.542 | |
| 1.079 |
| Dimethoxydiphenylsilane | |
| 1000 g | |
| MFCD00025690 | |
| 121°C (249°F) | |
| AHUXYBVKTIBBJW-UHFFFAOYSA-N | |
| dimethoxy(diphenyl)silane | |
| 81284 | |
| 97% | |
| Moisture sensitive |
Chemical Identifiers
| 6843-66-9 | |
| 244.365 | |
| AHUXYBVKTIBBJW-UHFFFAOYSA-N | |
| 81284 | |
| CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC |
| C14H16O2Si | |
| MFCD00025690 | |
| diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s | |
| dimethoxy(diphenyl)silane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 229-929-1
RTECSNumber : VV3643332
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only