missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydiphenylsilane, 97%

Catalog No. AAL0403118
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0403118 50 g
AAL0403130 250 g
AAL040310B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0403118 Supplier Thermo Scientific Chemicals Supplier No. L0403118
Only null left

CAS: 6843-66-9 | C14H16O2Si | 244.365 g/mol

Reagent for preparation of diphenylsilylene derivatives of diols by exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6843-66-9
Molecular Formula C14H16O2Si
Molecular Weight (g/mol) 244.365
MDL Number MFCD00025690
InChI Key AHUXYBVKTIBBJW-UHFFFAOYSA-N
Synonym diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s
PubChem CID 81284
IUPAC Name dimethoxy(diphenyl)silane
SMILES CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC

Specifications

Boiling Point 160°C to 162°C (15mmHg)
Quantity 50 g
Beilstein 2940458
Flash Point 121°C (249°F)
Formula Weight 244.37
Percent Purity 97%
Refractive Index 1.542
Sensitivity Moisture sensitive
Density 1.079
Chemical Name or Material Dimethoxydiphenylsilane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 229-929-1
RTECSNumber VV3643332
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only