missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimesitylfluoroborane 98.0+%, TCI America™
$100.11 - $306.10
Chemical Identifiers
| CAS | 436-59-9 |
|---|---|
| Molecular Formula | C18H22BF |
| Molecular Weight (g/mol) | 268.18 |
| MDL Number | MFCD00012203 |
| InChI Key | WZWGERGANZMXOM-UHFFFAOYSA-N |
| Synonym | Dimesitylboron Fluoride |
| PubChem CID | 3466040 |
| IUPAC Name | fluorobis(2,4,6-trimethylphenyl)borane |
| SMILES | CC1=CC(C)=C(B(F)C2=C(C)C=C(C)C=C2C)C(C)=C1 |
Chemical Identifiers
| 436-59-9 | |
| 268.18 | |
| WZWGERGANZMXOM-UHFFFAOYSA-N | |
| 3466040 | |
| CC1=CC(C)=C(B(F)C2=C(C)C=C(C)C=C2C)C(C)=C1 |
| C18H22BF | |
| MFCD00012203 | |
| Dimesitylboron Fluoride | |
| fluorobis(2,4,6-trimethylphenyl)borane |
Specifications
| 436-59-9 | |
| White-Yellow | |
| C18H22BF | |
| 1 g | |
| Dimesitylboron Fluoride | |
| CC1=CC(C)=C(B(F)C2=C(C)C=C(C)C=C2C)C(C)=C1 | |
| 268.18 | |
| 268.18 | |
| Crystalline Powder |
| 74°C | |
| 129°C | |
| MFCD00012203 | |
| 3261 | |
| WZWGERGANZMXOM-UHFFFAOYSA-N | |
| fluorobis(2,4,6-trimethylphenyl)borane | |
| 3466040 | |
| ≥98.0% (HPLC) | |
| Dimesitylfluoroborane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator