missing translation for 'onlineSavingsMsg'
Learn More

Dimenhydrinate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014449
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014449 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014449 Supplier Merck Emd Millipore Supplier No. PHR16581G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Dimenhydrinate, a diphenhydramine salt of 8-chlorotheophylline, exhibits antihistaminic activity, with antimuscarinic and sedative effects. It finds its major application as an antiemetic drug in the prevention and treatment of motion sickness. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 523-87-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H21 NO · C7H7 ClN4O2
MDL Number MFCD00054265
Synonym Dimenhydrinate; N-(2-Diphenylmethoxyethyl)-N,N-dimethylammonium 8-chlorotheophyllinate
Type Certified Reference Material
Quantity 1 g
Formula Weight 469.96
Grade Certified Reference Material