missing translation for 'onlineSavingsMsg'
Learn More

Dimedone 99.0+%, TCI America™

Catalog No. D070125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D070125G 25 g
D0701100G 100 g
D0701500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D070125G Supplier TCI America Supplier No. D070125G

Chemical Identifiers

CAS 126-81-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001588
InChI Key BADXJIPKFRBFOT-UHFFFAOYSA-N
Synonym dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
PubChem CID 31358
IUPAC Name 5,5-dimethylcyclohexane-1,3-dione
SMILES CC1(C)CC(=O)CC(=O)C1

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 25 g
Formula Weight 140.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimedone
EINECSNumber (3)-2374
RTECSNumber GV0345000
TSCA Yes