missing translation for 'onlineSavingsMsg'
Learn More

Interscience DiluFlow Booster Kit Dispensing Assembly

Catalog No. 16949056
Change view
Click to view available options
Quantity:
1 Kit (3.2 mm dia.)
1 Kit (6.4 mm dia.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16949056 1 Kit (6.4 mm dia.)
16949024 1 Kit (3.2 mm dia.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16949056 Supplier Interscience Supplier No. 513041

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Booster kit dispensing assembly for DiluFlow

Includes:

  • Set of silicone tubings, choose between inner dia. 3.2 mm or 6.4 mm
  • GL45 screw cap
  • Tube weight
  • Nozzle
  • Cap
  • Filter 'Y'

Specifications

Product Type Dispensing Tubing
Autoclavable Autoclavable
Quantity 1 Kit (6.4 mm dia.)
Sterility Non-sterile