missing translation for 'onlineSavingsMsg'
Learn More

Dilauryldimethylammonium Bromide 98.0+%, TCI America™

Catalog No. D1974500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1974500G 500 g
D197425G 25 g
D1974100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1974500G Supplier TCI America Supplier No. D1974500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3282-73-3
Molecular Formula C26H56BrN
Molecular Weight (g/mol) 462.645
MDL Number MFCD00041969
InChI Key XRWMGCFJVKDVMD-UHFFFAOYSA-M
Synonym didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1
PubChem CID 18669
IUPAC Name didodecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-]

Specifications

Melting Point 161°C
Color White
Quantity 500 g
Formula Weight 462.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dilauryldimethylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA Yes