missing translation for 'onlineSavingsMsg'
Learn More

Dilauroyl peroxide, 97%

CAS: 105-74-8 | C24H46O4 | 398.628 g/mol

$114.22 - $257.74

Chemical Identifiers

CAS 105-74-8
Molecular Formula C24H46O4
Molecular Weight (g/mol) 398.628
MDL Number MFCD00008964
InChI Key YIVJZNGAASQVEM-UHFFFAOYSA-N
Synonym lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle
PubChem CID 7773
ChEBI CHEBI:82479
IUPAC Name dodecanoyl dodecaneperoxoate
SMILES CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1431022
View Documents
Thermo Scientific Chemicals
L1431022
100 g
Each for $114.22
Only null left
 
AAL1431036
View Documents
Thermo Scientific Chemicals
L1431036
500 g
Each for $257.74
Only null left
 
Description

Description

Lauroyl peroxide acts as a bleaching agent, drying agent for fats, oils and waxes. Further, it serves as a polymerization initiator as well as vulcanizing agent. In addition to this, it plays an important role for high-pressure polyethylene and food used in bleaching agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lauroyl peroxide acts as a bleaching agent, drying agent for fats, oils and waxes. Further, it serves as a polymerization initiator as well as vulcanizing agent. In addition to this, it plays an important role for high-pressure polyethylene and food used in bleaching agent.

Solubility
Soluble in oils and organic solvents. Slightly soluble in alcohol. Insoluble in water.

Notes
Incompatible with acids, bases, metals, reducing agents, powdered metals, strong reducing agents and combustible material.
Specifications

Chemical Identifiers

105-74-8
398.628
YIVJZNGAASQVEM-UHFFFAOYSA-N
7773
dodecanoyl dodecaneperoxoate
C24H46O4
MFCD00008964
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle
CHEBI:82479
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC

Specifications

105-74-8
0.91
MFCD00008964
UN3106
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle
YIVJZNGAASQVEM-UHFFFAOYSA-N
dodecanoyl dodecaneperoxoate
7773
398.63
Lauroyl peroxide
53°C to 57°C
C24H46O4
100 g
1804936
Soluble in oils and organic solvents. Slightly soluble in alcohol. Insoluble in water.
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC
398.628
CHEBI:82479
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H242
Heating may cause a fire.

P210-P220-P234-P280-P501c

H242

DOTInformation : Hazard Class: 5.2

EINECSNumber : 203-326-3

RTECSNumber : OF2625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only