missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylphosphine, 10 wt.% solution in hexanes

Catalog No. AC383400100
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC383400100 10 g Glass bottle
AC383400025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC383400100 Supplier Thermo Scientific Chemicals Supplier No. 383400100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20491-53-6 | C6H15P | 118.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Diisopropylphosphine
CAS 20491-53-6, 110-54-3
Color Brown-Yellow to Yellow
Density 0.6700g/mL
Flash Point −22°C
Packaging Glass bottle
Molecular Formula C6H15P
Linear Formula [(CH3)2CH]2PH
Quantity 10 g
MDL Number MFCD00233814
Specific Gravity 0.67
Synonym diisopropylphosphine, di-isopropylphosphine, di-i-propylphosphine, diisopropylphosphane, phosphine, diisopropyl, phosphine, bis 1-methylethyl, ipr2ph, di-iso-propylphosphine, di-iso-propyl phosphine, di propan-2-yl phosphane
Solubility Information Solubility in water: insoluble
InChI Key WDIIYWASEVHBBT-UHFFFAOYSA-N
SMILES CC(C)PC(C)C
IUPAC Name di(propan-2-yl)phosphane
Molecular Weight (g/mol) 118.16
PubChem CID 536496
Formula Weight 118.16
Percent Purity 9 to 11% (Total base)
Physical Form Liquid
Show More Show Less

RUO – Research Use Only