missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropylphosphine, 10 wt.% solution in hexanes, AcroSeal™, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Diisopropylphosphine |
| CAS | 20491-53-6, 110-54-3 |
| Density | 0.6700g/mL |
| Flash Point | −22°C |
| Packaging | AcroSeal™ Glass bottle |
| Molecular Formula | C6H15P |
| Linear Formula | [(CH3)2CH]2PH |
| Quantity | 100mL |
| MDL Number | MFCD00233814 |
| Specific Gravity | 0.67 |
| Show More |
RUO – Research Use Only