missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropylcarbamoyl Chloride 98.0+%, TCI America™
$140.49 - $552.49
Chemical Identifiers
| CAS | 19009-39-3 |
|---|---|
| Molecular Formula | C7H14ClNO |
| Molecular Weight (g/mol) | 163.645 |
| MDL Number | MFCD00015013 |
| InChI Key | RSAFAYLZKCYUQW-UHFFFAOYSA-N |
| Synonym | diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride |
| PubChem CID | 87890 |
| IUPAC Name | N,N-di(propan-2-yl)carbamoyl chloride |
| SMILES | CC(C)N(C(C)C)C(=O)Cl |
Chemical Identifiers
| 19009-39-3 | |
| 163.645 | |
| RSAFAYLZKCYUQW-UHFFFAOYSA-N | |
| 87890 | |
| CC(C)N(C(C)C)C(=O)Cl |
| C7H14ClNO | |
| MFCD00015013 | |
| diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride | |
| N,N-di(propan-2-yl)carbamoyl chloride |
Specifications
| 59°C | |
| 100°C | |
| C7H14ClNO | |
| 3261 | |
| RSAFAYLZKCYUQW-UHFFFAOYSA-N | |
| N,N-di(propan-2-yl)carbamoyl chloride | |
| 163.645 | |
| 163.65 | |
| Crystalline Powder |
| 19009-39-3 | |
| White | |
| MFCD00015013 | |
| diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride | |
| CC(C)N(C(C)C)C(=O)Cl | |
| 5 g | |
| 87890 | |
| ≥98.0% (GC,T) | |
| Diisopropylcarbamoyl Chloride |
Safety and Handling
TSCA : No