missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylcarbamoyl Chloride 98.0+%, TCI America™

Catalog No. D41505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41505G 5 g
D415025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41505G Supplier TCI America Supplier No. D41505G

Chemical Identifiers

CAS 19009-39-3
Molecular Formula C7H14ClNO
Molecular Weight (g/mol) 163.645
MDL Number MFCD00015013
InChI Key RSAFAYLZKCYUQW-UHFFFAOYSA-N
Synonym diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride
PubChem CID 87890
IUPAC Name N,N-di(propan-2-yl)carbamoyl chloride
SMILES CC(C)N(C(C)C)C(=O)Cl

Specifications

Melting Point 59°C
Boiling Point 100°C
Color White
UN Number 3261
Quantity 5 g
Formula Weight 163.65
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diisopropylcarbamoyl Chloride