missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, 99+%, Thermo Scientific Chemicals

$73.48 - $73.48

Chemical Identifiers

CAS 108-18-9
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008862
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
PubChem CID 7912
SMILES CC(C)NC(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC219432500
SDS View Documents
Thermo Scientific Chemicals
219432500
250 mL Glass bottle
Each for $73.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-18-9
MFCD00008862
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
CC(C)NC(C)C
101.19
UAOMVDZJSHZZME-UHFFFAOYSA-N
7912

Specifications

108-18-9
0.7160g/mL
−17°C
99% min. (GC)
C6H15N
[(CH3)2CH]2NH
250 mL
0.716
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
UAOMVDZJSHZZME-UHFFFAOYSA-N
N-propan-2-ylpropan-2-amine
7912
101.19
Liquid
-61.0°C
84.0°C
Authentic
Glass bottle
1.3910 to 1.3930
MFCD00008862
04, 154
15, 3215
Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents
CC(C)NC(C)C
101.19
0.4 mPa.s (20°C)
99+%
Diisopropylamine, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

EINECSNumber : 203-558-5

RTECSNumber : IM4025000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only