missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl Phosphite 98.0+%, TCI America™
Supplier: TCI America P0629500G
Specifications
| Diisopropyl Phosphite | |
| Colorless | |
| C6H14O3P- | |
| 500 g | |
| IWEDBEZKWHRUJX-UHFFFAOYSA-N | |
| dipropan-2-yl phosphite | |
| 21884976 | |
| ≥98.0% (GC) |
| 1809-20-7 | |
| 79°C | |
| MFCD00117905 | |
| diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester | |
| CC(C)OP([O-])OC(C)C | |
| 165.149 | |
| 166.16 | |
| Liquid |
Chemical Identifiers
| 1809-20-7 | |
| 165.149 | |
| IWEDBEZKWHRUJX-UHFFFAOYSA-N | |
| 21884976 | |
| CC(C)OP([O-])OC(C)C |
| C6H14O3P- | |
| MFCD00117905 | |
| diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester | |
| dipropan-2-yl phosphite |
Safety and Handling
EINECSNumber : (2)-2001&(2)-1961
RTECSNumber : SZ7660000
TSCA : Yes