missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Phosphite 98.0+%, TCI America™

Catalog No. P062925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P062925G 25 g
P0629500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P062925G Supplier TCI America Supplier No. P062925G

Chemical Identifiers

CAS 1809-20-7
Molecular Formula C6H14O3P-
Molecular Weight (g/mol) 165.149
MDL Number MFCD00117905
InChI Key IWEDBEZKWHRUJX-UHFFFAOYSA-N
Synonym diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester
PubChem CID 21884976
IUPAC Name dipropan-2-yl phosphite
SMILES CC(C)OP([O-])OC(C)C

Specifications

Color Colorless
Boiling Point 79°C
Quantity 25 g
Formula Weight 166.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Phosphite
EINECSNumber (2)-2001&(2)-1961
RTECSNumber SZ7660000
TSCA Yes