missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl L-(+)-Tartrate 98.0+%, TCI America™
$52.19 - $508.89
Chemical Identifiers
| CAS | 2217-15-4 |
|---|---|
| Molecular Formula | C10H18O6 |
| Molecular Weight (g/mol) | 234.248 |
| MDL Number | MFCD00064450 |
| InChI Key | XEBCWEDRGPSHQH-HTQZYQBOSA-N |
| Synonym | +-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate |
| PubChem CID | 6453580 |
| IUPAC Name | dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate |
| SMILES | CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O |
Chemical Identifiers
| 2217-15-4 | |
| 234.248 | |
| XEBCWEDRGPSHQH-HTQZYQBOSA-N | |
| 6453580 | |
| CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O |
| C10H18O6 | |
| MFCD00064450 | |
| +-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate | |
| dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate |
Specifications
| 2217-15-4 | |
| 275°C | |
| MFCD00064450 | |
| +-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate | |
| CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O | |
| 234.248 | |
| 234.25 | |
| Liquid |
| Colorless | |
| C10H18O6 | |
| 25 g | |
| XEBCWEDRGPSHQH-HTQZYQBOSA-N | |
| dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate | |
| 6453580 | |
| ≥98.0% (GC) | |
| Diisopropyl L-(+)-Tartrate |
Safety and Handling
EINECSNumber : (2)-1458
RTECSNumber : WW8100000
TSCA : No