missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl L-(+)-Tartrate 98.0+%, TCI America™

Catalog No. T062125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T062125G 25 g
T0621500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T062125G Supplier TCI America Supplier No. T062125G

Chemical Identifiers

CAS 2217-15-4
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00064450
InChI Key XEBCWEDRGPSHQH-HTQZYQBOSA-N
Synonym +-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
PubChem CID 6453580
IUPAC Name dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate
SMILES CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O

Specifications

Color Colorless
Boiling Point 275°C
Quantity 25 g
Formula Weight 234.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl L-(+)-Tartrate
EINECSNumber (2)-1458
RTECSNumber WW8100000
TSCA No