missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Hydrazine-1,2-dicarboxylate 98.0+%, TCI America™

Catalog No. p-7128751
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5365200MG 200 mg
D53651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5365200MG Supplier TCI America Supplier No. D5365200MG

Chemical Identifiers

CAS 19740-72-8
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.23
MDL Number MFCD19443326
InChI Key FBZULTVJWVCJQV-UHFFFAOYSA-N
Synonym Hydrazine-1,2-dicarboxylic Acid Diisopropyl Ester
PubChem CID 88220
IUPAC Name N'-[(propan-2-yloxy)carbonyl](propan-2-yloxy)carbohydrazide
SMILES CC(C)OC(=O)NNC(=O)OC(C)C

Specifications

Melting Point 108°C
Color White
Quantity 200 mg
Formula Weight 204.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diisopropyl Hydrazine-1,2-dicarboxylate