missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ether, puriss p.a., ≥98.5% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120389
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Packaging:
Glass bottle
Catalog No. Quantity Packaging
60048427 1 L Glass bottle
1 options

Catalog No. 60048427

Supplier: Honeywell Inc 382701L

Only null left

Puriss. p.a.,≥98.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -86°C
Density 0.724g/mL at 20°C, 0.725g/mL at 25°C
Boiling Point 67°C to 70°C
Flash Point −29°C (−20.2°F)
Packaging Glass bottle
Refractive Index 1.367
Linear Formula (CH3)2CHOCH(CH3)2
Quantity 1 L
UN Number UN1159
Beilstein 1731256
Vapor Density 3.5 (vs air)
Formula Weight 102.17g/mol
Percent Purity ≥98.5% (GC)
Chemical Name or Material Diisopropyl ether
Show More Show Less
Hazard Statement P210-P261-P273
DOTInformation class 3 - PG 2 - Diisopropyl ether
RTECSNumber 203-560-6