missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl cyanomethylphosphonate, 97%

CAS: 21658-95-7 | C8H16NO3P | 205.194 g/mol

$149.52 - $575.49

Chemical Identifiers

CAS 21658-95-7
Molecular Formula C8H16NO3P
Molecular Weight (g/mol) 205.194
MDL Number MFCD00009812
InChI Key MQDIIJHQSUNBKA-UHFFFAOYSA-N
Synonym diisopropyl cyanomethyl phosphonate, diisopropyl cyanomethylphosphonate, diisopropyl cyanomethylphosphonate,, acmc-20al7r, diisopropoxyphosphinylacetonitrile, 2-di propan-2-yloxy phosphorylacetonitrile, diisopropyl cyanomethylphosphonate 5g, phosphonic acid,p-cyanomethyl-, bis 1-methylethyl ester
PubChem CID 88997
IUPAC Name 2-di(propan-2-yloxy)phosphorylacetonitrile
SMILES CC(C)OP(=O)(CC#N)OC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2216306
View Documents
Thermo Scientific Chemicals
B2216306
5 g
Each for $149.52
Only null left
 
AAB2216314
View Documents
Thermo Scientific Chemicals
B2216314
25 g
Each for $575.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21658-95-7
205.194
MQDIIJHQSUNBKA-UHFFFAOYSA-N
88997
CC(C)OP(=O)(CC#N)OC(C)C
C8H16NO3P
MFCD00009812
diisopropyl cyanomethyl phosphonate, diisopropyl cyanomethylphosphonate, diisopropyl cyanomethylphosphonate,, acmc-20al7r, diisopropoxyphosphinylacetonitrile, 2-di propan-2-yloxy phosphorylacetonitrile, diisopropyl cyanomethylphosphonate 5g, phosphonic acid,p-cyanomethyl-, bis 1-methylethyl ester
2-di(propan-2-yloxy)phosphorylacetonitrile

Specifications

21658-95-7
108°C to 110°C (0.08 mmHg)
C8H16NO3P
MFCD00009812
UN2927
diisopropyl cyanomethyl phosphonate, diisopropyl cyanomethylphosphonate, diisopropyl cyanomethylphosphonate,, acmc-20al7r, diisopropoxyphosphinylacetonitrile, 2-di propan-2-yloxy phosphorylacetonitrile, diisopropyl cyanomethylphosphonate 5g, phosphonic acid,p-cyanomethyl-, bis 1-methylethyl ester
CC(C)OP(=O)(CC#N)OC(C)C
205.194
205.19
Diisopropyl cyanomethylphosphonate
1.039
>110°C (230°F)
1.4295
5 g
1946176
MQDIIJHQSUNBKA-UHFFFAOYSA-N
2-di(propan-2-yloxy)phosphorylacetonitrile
88997
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 244-511-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only