missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl cyanomethylphosphonate, 97%

Catalog No. AAB2216314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2216314 25 g
AAB2216306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2216314 Supplier Thermo Scientific Chemicals Supplier No. B2216314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21658-95-7 | C8H16NO3P | 205.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21658-95-7
Molecular Formula C8H16NO3P
Molecular Weight (g/mol) 205.194
MDL Number MFCD00009812
InChI Key MQDIIJHQSUNBKA-UHFFFAOYSA-N
Synonym diisopropyl cyanomethyl phosphonate, diisopropyl cyanomethylphosphonate, diisopropyl cyanomethylphosphonate,, acmc-20al7r, diisopropoxyphosphinylacetonitrile, 2-di propan-2-yloxy phosphorylacetonitrile, diisopropyl cyanomethylphosphonate 5g, phosphonic acid,p-cyanomethyl-, bis 1-methylethyl ester
PubChem CID 88997
IUPAC Name 2-di(propan-2-yloxy)phosphorylacetonitrile
SMILES CC(C)OP(=O)(CC#N)OC(C)C

Specifications

Density 1.039
Boiling Point 108°C to 110°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4295
Quantity 25 g
UN Number UN2927
Beilstein 1946176
Formula Weight 205.19
Percent Purity 97%
Chemical Name or Material Diisopropyl cyanomethylphosphonate
Hazard Category H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 244-511-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only