missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Allylboronate 95.0+%, TCI America™

Catalog No. p-7127888
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37381G 1 g
D37385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37381G Supplier TCI America Supplier No. D37381G
Only null left

Chemical Identifiers

CAS 51851-79-7
Molecular Formula C9H19BO2
Molecular Weight (g/mol) 170.06
MDL Number MFCD15072157
InChI Key LWPLTONTMJTRJL-UHFFFAOYSA-N
Synonym Allylboronic Acid Diisopropyl Ester
PubChem CID 11435180
IUPAC Name bis(propan-2-yl) (prop-2-en-1-yl)boronate
SMILES CC(C)OB(CC=C)OC(C)C

Specifications

Color Colorless
Quantity 1 g
UN Number 1993
Formula Weight 170.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Allylboronate
TSCA No
Recommended Storage Refrigerator