missing translation for 'onlineSavingsMsg'
Learn More

Diisononyl Adipate (so called) [Plasticizer], TCI America™

$61.51 - $106.25

Chemical Identifiers

MDL Number MFCD00054165
Synonym Adipic Acid Diisononyl Ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A138725ML
View Documents
TCI America
A138725ML
25 mL
Each for $61.51
Only null left
 
A1387500ML
View Documents
TCI America
A1387500ML
500 mL
Each for $106.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00054165
Adipic Acid Diisononyl Ester

Specifications

Colorless
25 mL
Liquid
MFCD00054165
Adipic Acid Diisononyl Ester
Diisononyl Adipate (so called) [Plasticizer]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0861

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'