missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisodecyl 4-Cyclohexene-1,2-dicarboxylate 97.0+%, TCI America™
$78.29 - $508.89
Chemical Identifiers
| CAS | 87826-26-4 |
|---|---|
| Molecular Formula | C28H50O4 |
| Molecular Weight (g/mol) | 450.704 |
| MDL Number | MFCD00070615 |
| InChI Key | XRFYGUOAWVKOCQ-UHFFFAOYSA-N |
| Synonym | 4-Cyclohexene-1,2-dicarboxylic Acid Diisodecyl Ester, Diisodecyl 1,2,3,6-Tetrahydrophthalate, 1,2,3,6-Tetrahydrophthalic Acid Diisodecyl Ester |
| PubChem CID | 587973 |
| IUPAC Name | bis(8-methylnonyl) cyclohex-4-ene-1,2-dicarboxylate |
| SMILES | CC(C)CCCCCCCOC(=O)C1CC=CCC1C(=O)OCCCCCCCC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T060325ML
|
TCI America
T060325ML |
25 mL |
Each for $78.29
|
|
|||||
|
T0603500ML
|
TCI America
T0603500ML |
500 mL |
Each for $508.89
|
|
|||||
Chemical Identifiers
| 87826-26-4 | |
| 450.704 | |
| XRFYGUOAWVKOCQ-UHFFFAOYSA-N | |
| 587973 | |
| CC(C)CCCCCCCOC(=O)C1CC=CCC1C(=O)OCCCCCCCC(C)C |
| C28H50O4 | |
| MFCD00070615 | |
| 4-Cyclohexene-1,2-dicarboxylic Acid Diisodecyl Ester, Diisodecyl 1,2,3,6-Tetrahydrophthalate, 1,2,3,6-Tetrahydrophthalic Acid Diisodecyl Ester | |
| bis(8-methylnonyl) cyclohex-4-ene-1,2-dicarboxylate |
Specifications
| 87826-26-4 | |
| C28H50O4 | |
| 25 mL | |
| XRFYGUOAWVKOCQ-UHFFFAOYSA-N | |
| bis(8-methylnonyl) cyclohex-4-ene-1,2-dicarboxylate | |
| 587973 | |
| ≥97.0% (T) | |
| Diisodecyl 4-Cyclohexene-1,2-dicarboxylate |
| Colorless | |
| MFCD00070615 | |
| 4-Cyclohexene-1,2-dicarboxylic Acid Diisodecyl Ester, Diisodecyl 1,2,3,6-Tetrahydrophthalate, 1,2,3,6-Tetrahydrophthalic Acid Diisodecyl Ester | |
| CC(C)CCCCCCCOC(=O)C1CC=CCC1C(=O)OCCCCCCCC(C)C | |
| 450.704 | |
| 450.70 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2437
TSCA : No