missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylene (mixture of isomers) (stabilized with BHT) 94.0+%, TCI America™

Catalog No. p-7131234
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
I014725ML 25 mL
I0147500ML 500 mL
2 options

Catalog No. I014725ML

Supplier: TCI America I014725ML

Only null left

Chemical Identifiers

CAS 25167-70-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008855
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym Isooctene
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

Melting Point -106°C
Color Colorless
Boiling Point 105°C
Quantity 25 mL
UN Number 2050
Formula Weight 112.22
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutylene (mixture of isomers) (stabilized with BHT)
EINECSNumber (2)-0024
TSCA Yes