missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylene (mixture of isomers) (stabilized with BHT) 94.0+%, TCI America™

Catalog No. I0147500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0147500ML 500 mL
I014725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0147500ML Supplier TCI America Supplier No. I0147500ML
Only null left

Chemical Identifiers

CAS 25167-70-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008855
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym Isooctene
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

Melting Point -106°C
Color Colorless
Boiling Point 105°C
Quantity 500 mL
UN Number 2050
Formula Weight 112.22
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutylene (mixture of isomers) (stabilized with BHT)
EINECSNumber (2)-0024
TSCA Yes