missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylamine, 99%

Catalog No. p-7046637
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15495AP 500 mL
AAL15495AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15495AP Supplier Thermo Scientific Chemicals Supplier No. L15495AP
Only null left

CAS: 110-96-3 | C8H20N | 130.25 g/mol

Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.

Solubility
Soluble in water 5 g/L (20°C).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 110-96-3
Molecular Formula C8H20N
Molecular Weight (g/mol) 130.25
MDL Number MFCD00008930
InChI Key OBYVIBDTOCAXSN-OCAPTIKFSA-O
Synonym diisobutylamine, 1-propanamine, 2-methyl-n-2-methylpropyl, bis 2-methylpropyl amine, amine, diisobutyl, bis beta-methylpropyl amine, di-isobutylamine, n,n-bis 2-methylpropyl amine, unii-t18y0a819s, ccris 6232, di-2-methylpropyl amine
PubChem CID 8085
IUPAC Name 2-methyl-N-(2-methylpropyl)propan-1-amine
SMILES CC[C@H](C)[NH2+][C@H](C)CC

Specifications

Melting Point -77°C
Density 0.744
Boiling Point 137°C to 139°C
Flash Point 30°C (86°F)
Refractive Index 1.409
Quantity 500 mL
UN Number UN2361
Beilstein 1209251
Sensitivity Air Sensitive
Solubility Information Soluble in water 5g/L (20°C).
Formula Weight 129.25
Percent Purity 99%
Chemical Name or Material Diisobutylamine
Show More Show Less
Hazard Category H226-H301-H311-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIISOBUTYLAMINE
EINECSNumber 203-819-3
RTECSNumber TX1750000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only