missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Ether 98.0+%, TCI America™

Catalog No. D156525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D156525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D156525ML Supplier TCI America Supplier No. D156525ML
Only null left

Chemical Identifiers

CAS 628-55-7
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00008935
InChI Key SZNYYWIUQFZLLT-UHFFFAOYSA-N
Synonym Isobutyl Ether
PubChem CID 12346
IUPAC Name 2-methyl-1-(2-methylpropoxy)propane
SMILES CC(C)COCC(C)C

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 mL
UN Number 1149
Formula Weight 130.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutyl Ether
EINECSNumber (2)-0363
RTECSNumber NQ1000000
TSCA Yes